Product Details

SNP ID
rs12567084
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:227581028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAAGGTCATGTACAATTTTCA[C/T]TGGATAATTTTCAAAGTCTTCAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF678 PubMed Links

Gene Details

Gene
ZNF678
Gene Name
zinc finger protein 678
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178549.3 Intron NP_848644.2

View Full Product Details