Product Details

SNP ID
rs6577012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95305674 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCCTGCCTCCCCCCATCCTTAA[A/C]CCCTGGCCCTCTAATCTGTTCTCCA
Phenotype
MIM: 604662
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNIP3 PubMed Links

Gene Details

Gene
KCNIP3
Gene Name
potassium voltage-gated channel interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034914.1 Intron NP_001030086.1
NM_013434.4 Intron NP_038462.1

View Full Product Details