Product Details
- SNP ID
-
rs4595985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:3533924 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAGCCCGACCTTCAAGCCAAAGA[C/T]AGCCTGAAGCTGGAAAACCAAGCTG
- Phenotype
-
MIM: 604123
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEH1
PubMed Links
Gene Details
- Gene
- RNASEH1
- Gene Name
- ribonuclease H1
View Full Product Details