Product Details

SNP ID
rs5906244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46607844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGCGAGAGTGTCGGAGACGGCA[C/T]GAGGTCAGACAGGACCTTCTTCCTC
Phenotype
MIM: 300375 MIM: 300368
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
There are no transcripts associated with this gene.

Gene
SLC9A7
Gene Name
solute carrier family 9 member A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257291.1 Intron NP_001244220.1
NM_032591.2 Intron NP_115980.1
XM_005272681.4 Intron XP_005272738.1
XM_005272682.4 Intron XP_005272739.1
XM_006724564.3 Intron XP_006724627.1
XM_011543990.2 Intron XP_011542292.1
XM_017029905.1 Intron XP_016885394.1
XM_017029906.1 Intron XP_016885395.1

View Full Product Details