Product Details

SNP ID
rs11761037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1086899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAAAACGACACCTAGAAGTAGG[A/G]GTGAGATTCGCTGAAGTTCCCTTCT
Phenotype
MIM: 601805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf50 PubMed Links

Gene Details

Gene
C7orf50
Gene Name
chromosome 7 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134395.1 93 Intron NP_001127867.1
NM_001134396.1 93 Intron NP_001127868.1
NM_001318252.1 93 Intron NP_001305181.1
NM_032350.5 93 Intron NP_115726.1
XM_006715793.2 93 Intron XP_006715856.1
XM_011515580.2 93 Intron XP_011513882.1
XM_011515581.2 93 Intron XP_011513883.1
XM_011515582.2 93 Intron XP_011513884.1
XM_011515583.2 93 Intron XP_011513885.1
XM_011515584.2 93 Intron XP_011513886.1
XM_017012719.1 93 Intron XP_016868208.1
XM_017012720.1 93 Intron XP_016868209.1
XM_017012721.1 93 Intron XP_016868210.1
XM_017012722.1 93 Intron XP_016868211.1
Gene
GPER1
Gene Name
G protein-coupled estrogen receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039966.1 93 UTR 5 NP_001035055.1
NM_001098201.1 93 Intron NP_001091671.1
NM_001505.2 93 UTR 5 NP_001496.1

View Full Product Details