Product Details
- SNP ID
-
rs4075258
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48525876 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTCAGTCATTGTGCCCAAGATGC[T/C]GGAGAACCTCCTGTCACAGAGGAAA
- Phenotype
-
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR8S1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35367885] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR8S1
- Gene Name
- olfactory receptor family 8 subfamily S member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005203.2 |
245 |
Missense Mutation |
CCG,CTG |
P82L |
NP_001005203.2 |
View Full Product Details