Product Details
- SNP ID
-
rs6593674
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:95103445 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTATGTTAAGTTGGGGAGAATC[A/G]TGAAATGTTTAGAGATAATTTCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM56
PubMed Links
Gene Details
- Gene
- TMEM56
- Gene Name
- transmembrane protein 56
View Full Product Details