Product Details
- SNP ID
-
rs4788055
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28271966 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTACCCAAATAAGTTGAAAATTTA[C/T]GCCCACAGAAAATCCTGCACATGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SBK1
PubMed Links
Gene Details
- Gene
- SBK1
- Gene Name
- SH3 domain binding kinase 1
View Full Product Details