Product Details

SNP ID
rs11099554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82819714 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTATAAAACCTGTAGTTTTTT[A/C]CTATGTCATTACTTGGCATAGGTGT
Phenotype
MIM: 610257
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC31A PubMed Links

Gene Details

Gene
SEC31A
Gene Name
SEC31 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077206.3 Intron NP_001070674.1
NM_001077207.3 Intron NP_001070675.1
NM_001077208.3 Intron NP_001070676.1
NM_001191049.2 Intron NP_001177978.1
NM_001300744.2 Intron NP_001287673.1
NM_001300745.2 Intron NP_001287674.1
NM_001318119.1 Intron NP_001305048.1
NM_001318120.1 Intron NP_001305049.1
NM_016211.4 Intron NP_057295.2

View Full Product Details