Product Details

SNP ID
rs4689519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4423893 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGCATTCTGTGTCACTGCTCGG[A/G]GAATGGGGAGGGCCTGGTTGTGCCC
Phenotype
MIM: 607645 MIM: 606046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSG1 PubMed Links

Gene Details

Gene
NSG1
Gene Name
neuron specific gene family member 1
There are no transcripts associated with this gene.

Gene
STX18
Gene Name
syntaxin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016930.2 Intron NP_058626.1
XM_011513480.1 Intron XP_011511782.1
XM_011513483.1 Intron XP_011511785.1
XM_011513485.2 Intron XP_011511787.1
XM_017008296.1 Intron XP_016863785.1
XM_017008297.1 Intron XP_016863786.1
XM_017008298.1 Intron XP_016863787.1

View Full Product Details