Product Details

SNP ID
rs10282065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43609649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCAAGTCCAGGTGCAACTGTGGG[A/G]AAATGAAACATTCCTCAAACACAGA
Phenotype
MIM: 614769 MIM: 604726
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COA1 PubMed Links

Gene Details

Gene
COA1
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321197.1 Intron NP_001308126.1
NM_001321198.1 Intron NP_001308127.1
NM_001321199.1 Intron NP_001308128.1
NM_001321200.1 Intron NP_001308129.1
NM_001321201.1 Intron NP_001308130.1
NM_001321202.1 Intron NP_001308131.1
NM_001321203.1 Intron NP_001308132.1
NM_001321204.1 Intron NP_001308133.1
NM_001321205.1 Intron NP_001308134.1
NM_018224.3 Intron NP_060694.2
XM_005249807.4 Intron XP_005249864.1
XM_006715752.2 Intron XP_006715815.1
XM_017012413.1 Intron XP_016867902.1
XM_017012414.1 Intron XP_016867903.1
XM_017012415.1 Intron XP_016867904.1
XM_017012416.1 Intron XP_016867905.1
XM_017012417.1 Intron XP_016867906.1
XM_017012418.1 Intron XP_016867907.1
XM_017012419.1 Intron XP_016867908.1
XM_017012420.1 Intron XP_016867909.1
XM_017012421.1 Intron XP_016867910.1
XM_017012422.1 Intron XP_016867911.1
XM_017012423.1 Intron XP_016867912.1
XM_017012424.1 Intron XP_016867913.1
XM_017012425.1 Intron XP_016867914.1
XM_017012426.1 Intron XP_016867915.1
XM_017012427.1 Intron XP_016867916.1
XM_017012428.1 Intron XP_016867917.1
XM_017012429.1 Intron XP_016867918.1
XM_017012430.1 Intron XP_016867919.1
XM_017012431.1 Intron XP_016867920.1
XM_017012432.1 Intron XP_016867921.1
Gene
STK17A
Gene Name
serine/threonine kinase 17a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004760.2 Intron NP_004751.2
XM_017012792.1 Intron XP_016868281.1

View Full Product Details