Product Details
- SNP ID
-
rs11534413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137427214 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTTTCAAGCACAAATTCATGCA[C/T]GCTAGCGACAGGCTCACGGGTGTGA
- Phenotype
-
MIM: 616748
MIM: 611255
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ENTPD8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11522288,rs11534414] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ENTPD8
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 8
There are no transcripts associated with this gene.
- Gene
- EXD3
- Gene Name
- exonuclease 3'-5' domain containing 3
There are no transcripts associated with this gene.
- Gene
- NOXA1
- Gene Name
- NADPH oxidase activator 1
View Full Product Details