Product Details
- SNP ID
-
rs12471385
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:95141224 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAGCATTACAAGAACAGGCTAATA[C/T]AGGAACCACATAACAAAACTCCATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF514
PubMed Links
Gene Details
- Gene
- ZNF514
- Gene Name
- zinc finger protein 514
View Full Product Details