Product Details

SNP ID
rs11133442
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:56440301 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCTGTATCTTAAAGGGACTTTTAA[A/T]CCATGTTAATGAGTCTAGACATTAC
Phenotype
MIM: 172439 MIM: 172450
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PAICS PubMed Links

Gene Details

Gene
PAICS
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079524.1 Intron NP_001072992.1
NM_001079525.1 Intron NP_001072993.1
NM_006452.3 Intron NP_006443.1
XM_011534366.1 Intron XP_011532668.1
Gene
PPAT
Gene Name
phosphoribosyl pyrophosphate amidotransferase
There are no transcripts associated with this gene.

View Full Product Details