Product Details

SNP ID
rs35488977
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:42791093 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAACAAAATACATACAGTAGTTGAG[G/T]AACAAATCTCTCATATGTTTAGTAT
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HIGD1A PubMed Links
Additional Information
For this assay, SNP(s) [rs73075301] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HIGD1A
Gene Name
HIG1 hypoxia inducible domain family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099668.1 Intron NP_001093138.1
NM_001099669.1 Intron NP_001093139.1
NM_014056.3 Intron NP_054775.2

View Full Product Details