Product Details

SNP ID
rs126014
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:28893666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGCAAGACCCCAGAGGCAGAGG[A/G]AGGGCAGGGTTCCCTCCTGGCTCAG
Phenotype
MIM: 130500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166005.1 312 Intron NP_001159477.1
NM_001166006.1 312 Intron NP_001159478.1
NM_001166007.1 312 Intron NP_001159479.1
NM_004437.3 312 Intron NP_004428.1
NM_203342.2 312 Intron NP_976217.1
NM_203343.2 312 Intron NP_976218.1
XM_005245753.1 312 Intron XP_005245810.1
XM_005245757.1 312 Intron XP_005245814.1
XM_005245760.1 312 Intron XP_005245817.1
XM_005245761.1 312 Intron XP_005245818.1
XM_005245763.1 312 Intron XP_005245820.1
XM_005245764.1 312 Intron XP_005245821.1
XM_005245765.1 312 Intron XP_005245822.1
XM_005245768.1 312 Intron XP_005245825.1
XM_005245769.1 312 Intron XP_005245826.1
XM_005245770.1 312 Intron XP_005245827.1
XM_005245772.4 312 Intron XP_005245829.1
XM_005245773.4 312 Intron XP_005245830.1
XM_005245774.1 312 Intron XP_005245831.1
XM_006710434.1 312 Intron XP_006710497.1
XM_006710439.1 312 Intron XP_006710502.1
XM_011540956.1 312 Intron XP_011539258.1
XM_011540957.1 312 Intron XP_011539259.1
XM_011540958.1 312 Intron XP_011539260.1
XM_011540959.1 312 Intron XP_011539261.1
XM_011540960.1 312 Intron XP_011539262.1
XM_011540961.1 312 Intron XP_011539263.1
XM_011540962.1 312 Intron XP_011539264.1
XM_011540963.2 312 Intron XP_011539265.1
XM_011540964.1 312 Intron XP_011539266.1
XM_011540965.2 312 Intron XP_011539267.1
XM_017000581.1 312 Intron XP_016856070.1
XM_017000582.1 312 Intron XP_016856071.1
XM_017000583.1 312 Intron XP_016856072.1
XM_017000584.1 312 UTR 5 XP_016856073.1
XM_017000585.1 312 UTR 5 XP_016856074.1
XM_017000586.1 312 Intron XP_016856075.1
XM_017000587.1 312 Intron XP_016856076.1
XM_017000588.1 312 Intron XP_016856077.1
XM_017000589.1 312 Intron XP_016856078.1
XM_017000590.1 312 Intron XP_016856079.1
XM_017000591.1 312 Intron XP_016856080.1
XM_017000592.1 312 Intron XP_016856081.1
XM_017000593.1 312 Intron XP_016856082.1
XM_017000594.1 312 Intron XP_016856083.1
XM_017000595.1 312 Intron XP_016856084.1
XM_017000596.1 312 Intron XP_016856085.1
XM_017000597.1 312 Intron XP_016856086.1
XM_017000598.1 312 Intron XP_016856087.1
XM_017000599.1 312 Intron XP_016856088.1
XM_017000600.1 312 Intron XP_016856089.1
XM_017000601.1 312 Intron XP_016856090.1
XM_017000602.1 312 Intron XP_016856091.1
XM_017000603.1 312 Intron XP_016856092.1
XM_017000604.1 312 Intron XP_016856093.1

View Full Product Details