Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001037329.3 | Intron | NP_001032406.1 | |||
XM_017017217.1 | Intron | XP_016872706.1 | |||
XM_017017218.1 | Intron | XP_016872707.1 | |||
XM_017017219.1 | Intron | XP_016872708.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098794.1 | Intron | NP_001092264.1 | |||
NM_032127.3 | Intron | NP_115503.2 | |||
XM_006718343.2 | Intron | XP_006718406.1 | |||
XM_011520397.2 | Intron | XP_011518699.1 | |||
XM_011520398.2 | Intron | XP_011518700.1 | |||
XM_011520399.2 | Intron | XP_011518701.1 |