Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040665.1 | 2991 | Intron | NP_001035755.1 | ||
NM_001040666.1 | 2991 | Intron | NP_001035756.1 | ||
NM_001244944.1 | 2991 | Intron | NP_001231873.1 | ||
NM_001244945.1 | 2991 | Intron | NP_001231874.1 | ||
NM_001244946.1 | 2991 | Intron | NP_001231875.1 | ||
NM_152999.3 | 2991 | Intron | NP_694544.2 | ||
XM_006715921.3 | 2991 | Intron | XP_006715984.1 | ||
XM_017011953.1 | 2991 | Intron | XP_016867442.1 | ||
XM_017011954.1 | 2991 | Intron | XP_016867443.1 | ||
XM_017011955.1 | 2991 | Intron | XP_016867444.1 | ||
XM_017011956.1 | 2991 | Intron | XP_016867445.1 | ||
XM_017011957.1 | 2991 | Intron | XP_016867446.1 | ||
XM_017011958.1 | 2991 | UTR 5 | XP_016867447.1 |