Product Details

SNP ID
rs222493
Assay Type
Validated
NCBI dbSNP Submissions
70
Location
Chr.1:114585466 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAGCAACAGGAGAATCCTCCTCT[A/G]ACAGCCTGACTCGCTCTTTAACCTT
Phenotype
MIM: 605783
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
There are no transcripts associated with this gene.

Gene
DENND2C
Gene Name
DENN domain containing 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256404.1 3623 UTR 3 NP_001243333.1
NM_198459.3 3623 UTR 3 NP_940861.3

View Full Product Details