Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004160.5 | Intron | NP_004151.3 | |||
XM_011525035.1 | Intron | XP_011523337.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001304813.1 | Intron | NP_001291742.1 | |||
NM_001304814.1 | Intron | NP_001291743.1 | |||
NM_032376.3 | Intron | NP_115752.1 | |||
XM_005257737.2 | Intron | XP_005257794.1 | |||
XM_011525346.1 | Intron | XP_011523648.1 | |||
XM_011525347.1 | Intron | XP_011523649.1 | |||
XM_011525349.1 | Intron | XP_011523651.1 | |||
XM_011525351.1 | Intron | XP_011523653.1 | |||
XM_011525353.1 | Intron | XP_011523655.1 |