Product Details
- SNP ID
-
rs423561
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:48562713 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTATGCCCAGATGTGGCAGCATCC[A/G]TCCAGCTGTGTGGCTGCCCTCCAGG
- Phenotype
-
MIM: 120120
MIM: 605320
MIM: 605902
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COL7A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78159609] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL7A1
- Gene Name
- collagen type VII alpha 1
There are no transcripts associated with this gene.
- Gene
- PFKFB4
- Gene Name
- 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4
- Gene
- UCN2
- Gene Name
- urocortin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033199.3 |
490 |
UTR 3 |
|
|
NP_149976.1 |
View Full Product Details