Product Details
- SNP ID
-
rs211578
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:159177599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAATCAGGAGAAGCTGCTTTTGA[C/T]AAAAGCTCTGTGAAATTTATGGGTG
- Phenotype
-
MIM: 609991
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FNDC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141031566] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FNDC1
- Gene Name
- fibronectin type III domain containing 1
- Gene
- LOC101929122
- Gene Name
- uncharacterized LOC101929122
There are no transcripts associated with this gene.
View Full Product Details