Product Details

SNP ID
rs531651
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:20500538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGCAGCATCCTGCCATTGGAGGC[A/G]TGGGTCTGGAGAGTTTCTACCCAAT
Phenotype
MIM: 612037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUL1 PubMed Links

Gene Details

Gene
MUL1
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024544.2 2051 UTR 3 NP_078820.2
XM_011542137.2 2051 UTR 3 XP_011540439.1

View Full Product Details