Product Details

SNP ID
rs229665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64704600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCGGGAGCGGTCGGCGGCCTC[A/G]GCCCCTTCAGAGAGCGACTTTCAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG3 PubMed Links

Gene Details

Gene
PLEKHG3
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308147.1 472 UTR 5 NP_001295076.1
XM_011536624.2 472 Intron XP_011534926.1
XM_011536625.2 472 Intron XP_011534927.1
XM_011536627.2 472 Intron XP_011534929.1
XM_011536628.2 472 UTR 5 XP_011534930.1
XM_011536629.2 472 Intron XP_011534931.1
XM_017021154.1 472 Intron XP_016876643.1
XM_017021155.1 472 Intron XP_016876644.1
XM_017021156.1 472 Intron XP_016876645.1
XM_017021157.1 472 Intron XP_016876646.1
XM_017021158.1 472 Intron XP_016876647.1

View Full Product Details