Product Details

SNP ID
rs30461
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:39298475 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGACCTCAAATATGTGGCCGATGGG[A/G]ACCTGTGTCTGAGAACGTCAACCCA
Phenotype
MIM: 607403 MIM: 612807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNL1 PubMed Links

Gene Details

Gene
IFNL1
Gene Name
interferon lambda 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172140.1 659 Missense Mutation AAC,GAC N188D NP_742152.1
Gene
LRFN1
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
There are no transcripts associated with this gene.

View Full Product Details