Product Details

SNP ID
rs4549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42594649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATGAAACCTCTGTGGCTCTTTC[A/G]AAACGTGAAAATGTGAAGAAAGCTA
Phenotype
MIM: 161565 MIM: 604028 MIM: 606473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKTR PubMed Links

Gene Details

Gene
NKTR
Gene Name
natural killer cell triggering receptor
There are no transcripts associated with this gene.

Gene
SEC22C
Gene Name
SEC22 homolog C, vesicle trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201572.1 1594 Intron NP_001188501.1
NM_001201584.1 1594 Intron NP_001188513.1
NM_004206.3 1594 Intron NP_004197.1
NM_032970.3 1594 Intron NP_116752.1
XM_005265533.1 1594 Intron XP_005265590.1
XM_005265534.1 1594 Intron XP_005265591.1
XM_005265535.1 1594 Intron XP_005265592.1
XM_011534212.1 1594 Intron XP_011532514.1
XM_017007438.1 1594 Intron XP_016862927.1
XM_017007439.1 1594 Intron XP_016862928.1
Gene
SS18L2
Gene Name
SS18 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016305.3 1594 Intron NP_057389.1
XM_017006548.1 1594 UTR 3 XP_016862037.1
XM_017006549.1 1594 Intron XP_016862038.1

View Full Product Details