Product Details

SNP ID
rs642199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28654389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACACTGCAAGTTGGAAAACAT[G/T]GGGTTTCTTTATTGCAGAATATCAG
Phenotype
MIM: 610664 MIM: 602934 MIM: 601333
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0100 PubMed Links

Gene Details

Gene
KIAA0100
Gene Name
KIAA0100
There are no transcripts associated with this gene.

Gene
SDF2
Gene Name
stromal cell derived factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006923.3 Intron NP_008854.2
XM_011525106.2 Intron XP_011523408.1
Gene
SUPT6H
Gene Name
SPT6 homolog, histone chaperone
There are no transcripts associated with this gene.

View Full Product Details