Product Details

SNP ID
rs536455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57387528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGGCAGTGGGCTCGACGCCAGT[A/G]GCCTCCTGTGAAGGCAGAACAGAAA
Phenotype
MIM: 605601 MIM: 606814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRG2 PubMed Links

Gene Details

Gene
PRG2
Gene Name
proteoglycan 2, pro eosinophil major basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243245.2 803 Missense Mutation CAC,TAC H195Y NP_001230174.1
NM_001302926.1 803 Missense Mutation CAC,TAC H206Y NP_001289855.1
NM_001302927.1 803 Missense Mutation CAC,TAC H206Y NP_001289856.1
NM_002728.5 803 Missense Mutation CAC,TAC H206Y NP_002719.3
Gene
PRG3
Gene Name
proteoglycan 3, pro eosinophil major basic protein 2
There are no transcripts associated with this gene.

View Full Product Details