Product Details

SNP ID
rs553074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31382425 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGATGCCTTATCATAATTTATCAT[C/T]ATCAGTAGAAATTATGTACAAGATG
Phenotype
MIM: 611072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC24 PubMed Links

Gene Details

Gene
DNAJC24
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181706.4 Intron NP_859057.4
XM_017017216.1 Intron XP_016872705.1

View Full Product Details