Product Details

SNP ID
rs612888
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:31375309 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACTACCTCACCTGATTCTCACTCA[C/T]TGTTTCAAAATGATCTTAGTTGTAA
Phenotype
MIM: 608062 MIM: 611072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCDC1 PubMed Links

Gene Details

Gene
DCDC1
Gene Name
doublecortin domain containing 1
There are no transcripts associated with this gene.

Gene
DNAJC24
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181706.4 Intron NP_859057.4
XM_017017216.1 Intron XP_016872705.1

View Full Product Details