Product Details

SNP ID
rs442366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9793779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACATGTGGGCCTTTTTCATAC[A/G]TGTGGGCATTCCTGGGAGTTGTGAC
Phenotype
MIM: 604226 MIM: 601982 MIM: 602945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC4 PubMed Links

Gene Details

Gene
ARPC4
Gene Name
actin related protein 2/3 complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024959.2 Intron NP_001020130.1
NM_001024960.2 Intron NP_001020131.1
NM_001198780.1 Intron NP_001185709.1
NM_005718.4 Intron NP_005709.1
Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198793.1 Intron NP_001185722.1
Gene
OGG1
Gene Name
8-oxoguanine DNA glycosylase
There are no transcripts associated with this gene.

Gene
TADA3
Gene Name
transcriptional adaptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278270.1 Intron NP_001265199.1
NM_006354.3 Intron NP_006345.1
NM_133480.2 Intron NP_597814.1

View Full Product Details