Product Details

SNP ID
rs678132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124623303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACAAAATCTTTCCCCAAGCTGTT[A/C]CCCCTTCCCAGTGTGACAGTATTAA
Phenotype
MIM: 608422 MIM: 610614
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PANX3 PubMed Links

Gene Details

Gene
PANX3
Gene Name
pannexin 3
There are no transcripts associated with this gene.

Gene
TBRG1
Gene Name
transforming growth factor beta regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032811.2 Intron NP_116200.2
XM_005271709.3 Intron XP_005271766.1
XM_017018442.1 Intron XP_016873931.1

View Full Product Details