Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173551.4 | 3672 | UTR 3 | NP_775822.3 | ||
XM_005251793.4 | 3672 | Intron | XP_005251850.1 | ||
XM_005251794.4 | 3672 | Intron | XP_005251851.1 | ||
XM_006716998.3 | 3672 | Intron | XP_006717061.1 | ||
XM_006716999.2 | 3672 | Intron | XP_006717062.1 | ||
XM_011518357.2 | 3672 | Intron | XP_011516659.1 | ||
XM_017014445.1 | 3672 | Intron | XP_016869934.1 | ||
XM_017014446.1 | 3672 | Intron | XP_016869935.1 | ||
XM_017014447.1 | 3672 | Intron | XP_016869936.1 |