Product Details

SNP ID
rs184400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57431939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATCTTTTCAGGTTTACACATG[C/G]CCACAAGAGTTTTCAACATATCGTA
Phenotype
MIM: 608943
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIAPIN1 PubMed Links

Gene Details

Gene
CIAPIN1
Gene Name
cytokine induced apoptosis inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308347.1 Intron NP_001295276.1
NM_001308358.1 Intron NP_001295287.1
NM_020313.3 Intron NP_064709.2
XM_005256063.4 Intron XP_005256120.1

View Full Product Details