Product Details

SNP ID
rs478776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139045869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAATAAGCATTTGTAGTAACT[C/T]TCCCAGAAAATAACTCTACTCTAAA
Phenotype
MIM: 607312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3HAV1 PubMed Links

Gene Details

Gene
ZC3HAV1
Gene Name
zinc finger CCCH-type containing, antiviral 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020119.3 4822 UTR 3 NP_064504.2
NM_024625.3 4822 Intron NP_078901.3
XM_005250501.2 4822 UTR 3 XP_005250558.1
XM_011516412.2 4822 Intron XP_011514714.1
Gene
ZC3HAV1L
Gene Name
zinc finger CCCH-type containing, antiviral 1 like
There are no transcripts associated with this gene.

View Full Product Details