Product Details

SNP ID
rs640273
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:99647348 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGTAATACATACTGAATGTCACT[A/C]TAAGATATGTAGAGAGATTTCTAAA
Phenotype
MIM: 120251
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
COL8A1 PubMed Links

Gene Details

Gene
COL8A1
Gene Name
collagen type VIII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001850.4 Intron NP_001841.2
NM_020351.3 Intron NP_065084.2

View Full Product Details