Product Details

SNP ID
rs651157
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143339898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAGAAACTAGGATTTTCTTTGC[C/T]TGGTGTAGCACTGGTGTTTTCTTTA
Phenotype
MIM: 604439
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GAB1 PubMed Links

Gene Details

Gene
GAB1
Gene Name
GRB2 associated binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002039.3 Intron NP_002030.2
NM_207123.2 Intron NP_997006.1
XM_006714167.3 Intron XP_006714230.1
XM_006714168.3 Intron XP_006714231.1
XM_017007966.1 Intron XP_016863455.1
XM_017007967.1 Intron XP_016863456.1
XM_017007968.1 Intron XP_016863457.1
XM_017007969.1 Intron XP_016863458.1
XM_017007970.1 Intron XP_016863459.1
XM_017007971.1 Intron XP_016863460.1
Gene
MIR3139
Gene Name
microRNA 3139
There are no transcripts associated with this gene.

View Full Product Details