Product Details
- SNP ID
-
rs6039
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113105914 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGGCTGCCTGGCTGCAGGTGCGTCC[A/G]GGGAGGTTTTCTCCATAAACTTGGT
- Phenotype
-
MIM: 613878
MIM: 609499
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
F7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10482844] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- F7
- Gene Name
- coagulation factor VII
- Gene
- MCF2L
- Gene Name
- MCF.2 cell line derived transforming sequence like
There are no transcripts associated with this gene.
View Full Product Details