Product Details

SNP ID
rs732253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183710439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACATTACACATTATTATTCTTG[G/T]GAGACAGAAGCAGACCTGGATTATT
Phenotype
MIM: 613373
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YEATS2 PubMed Links

Gene Details

Gene
YEATS2
Gene Name
YEATS domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018023.4 Intron NP_060493.3

View Full Product Details