Product Details

SNP ID
rs728116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16501167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTGACTCAATGACCGCCGATGC[C/T]GGTCTTCCAAAGAGAGCTGACAGAA
Phenotype
MIM: 611414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf44 PubMed Links

Gene Details

Gene
C19orf44
Gene Name
chromosome 19 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288834.1 531 Silent Mutation GCC,GCT A125A NP_001275763.1
NM_032207.3 531 Silent Mutation GCC,GCT A125A NP_115583.1
XM_006722920.3 531 Silent Mutation GCC,GCT A125A XP_006722983.1
XM_011528354.2 531 Silent Mutation GCC,GCT A125A XP_011526656.1
XM_017027350.1 531 Silent Mutation GCC,GCT A125A XP_016882839.1
Gene
CALR3
Gene Name
calreticulin 3
There are no transcripts associated with this gene.

View Full Product Details