Product Details

SNP ID
rs749621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109441397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAAGCACCGGGCCGACCGCATG[A/C]GGCGCCTGGGAGACTTTCTGAACCA
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
There are no transcripts associated with this gene.

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111298.2 485 Intron NP_001104768.2
NM_001286360.1 485 Intron NP_001273289.1
NM_001286361.1 485 Intron NP_001273290.1
NM_173672.4 485 Intron NP_775943.1
XM_011535765.2 485 Intron XP_011534067.1
XM_011535766.2 485 Intron XP_011534068.1
XM_011535767.2 485 Intron XP_011534069.1
XM_011535769.2 485 Intron XP_011534071.1
XM_017010774.1 485 Intron XP_016866263.1
Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 485 Silent Mutation AGG,CGG R31R NP_003071.2
XM_005267109.1 485 UTR 5 XP_005267166.1
XM_011536079.1 485 Intron XP_011534381.1

View Full Product Details