Product Details

SNP ID
rs324091
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:52401814 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAACATCTGCTTGCTCAGTTAGG[A/C]TAAATCAGACCTATGTAGAAAGTCA
Phenotype
MIM: 615580
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF528 PubMed Links

Gene Details

Gene
ZNF528
Gene Name
zinc finger protein 528
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032423.2 Intron NP_115799.2
XM_005259323.1 Intron XP_005259380.1
XM_006723418.2 Intron XP_006723481.1
XM_011527392.1 Intron XP_011525694.1
XM_011527394.2 Intron XP_011525696.1
XM_017027360.1 Intron XP_016882849.1
XM_017027361.1 Intron XP_016882850.1
XM_017027362.1 Intron XP_016882851.1
XM_017027363.1 Intron XP_016882852.1
XM_017027364.1 Intron XP_016882853.1
XM_017027365.1 Intron XP_016882854.1
Gene
ZNF528-AS1
Gene Name
ZNF528 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF880
Gene Name
zinc finger protein 880
There are no transcripts associated with this gene.

View Full Product Details