Product Details

SNP ID
rs429216
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78130123 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGACAGAGAATTGGCAAAGATA[G/T]GTCTTAAACATCTAGCCTGGGAGAG
Phenotype
MIM: 605828 MIM: 605829
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TMC6 PubMed Links

Gene Details

Gene
TMC6
Gene Name
transmembrane channel like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127198.2 828 Intron NP_001120670.1
NM_001321185.1 828 Intron NP_001308114.1
NM_007267.7 828 Intron NP_009198.4
XM_005256995.1 828 Intron XP_005257052.1
XM_011524255.1 828 Intron XP_011522557.1
XM_011524256.1 828 Intron XP_011522558.1
XM_011524257.2 828 Intron XP_011522559.1
XM_011524258.1 828 Intron XP_011522560.1
XM_017024107.1 828 Intron XP_016879596.1
XM_017024108.1 828 Intron XP_016879597.1
XM_017024109.1 828 Intron XP_016879598.1
Gene
TMC8
Gene Name
transmembrane channel like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152468.4 828 Intron NP_689681.2
XM_011524402.2 828 UTR 5 XP_011522704.1
XM_011524403.2 828 UTR 5 XP_011522705.1
XM_011524404.2 828 UTR 5 XP_011522706.1
XM_011524406.2 828 UTR 5 XP_011522708.1
XM_011524409.2 828 UTR 5 XP_011522711.1
XM_011524410.2 828 UTR 5 XP_011522712.1
XM_011524411.2 828 UTR 5 XP_011522713.1
XM_017024238.1 828 UTR 5 XP_016879727.1
XM_017024239.1 828 UTR 5 XP_016879728.1
XM_017024240.1 828 UTR 5 XP_016879729.1
XM_017024241.1 828 UTR 5 XP_016879730.1
XM_017024242.1 828 Intron XP_016879731.1
XM_017024243.1 828 Intron XP_016879732.1
XM_017024244.1 828 UTR 5 XP_016879733.1

View Full Product Details