Product Details

SNP ID
rs182662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57189210 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCATTTTGTAAAAGAAATGATT[C/T]TGTATGTGGGACTGACAGCTCTTGA
Phenotype
MIM: 603882 MIM: 606144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG2 PubMed Links

Gene Details

Gene
BAG2
Gene Name
BCL2 associated athanogene 2
There are no transcripts associated with this gene.

Gene
RAB23
Gene Name
RAB23, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278666.1 2173 UTR 3 NP_001265595.1
NM_001278667.1 2173 UTR 3 NP_001265596.1
NM_001278668.1 2173 UTR 3 NP_001265597.1
NM_016277.4 2173 UTR 3 NP_057361.3
NM_183227.2 2173 UTR 3 NP_899050.1

View Full Product Details