Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_153371.3 | 170 | Intron | NP_699202.1 | ||
XM_011534995.2 | 170 | Intron | XP_011533297.1 | ||
XM_017020434.1 | 170 | Intron | XP_016875923.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001206559.1 | 170 | UTR 5 | NP_001193488.1 | ||
NM_015972.3 | 170 | Intron | NP_057056.1 | ||
NM_152705.2 | 170 | Intron | NP_689918.1 | ||
XM_005266412.1 | 170 | UTR 5 | XP_005266469.1 | ||
XM_005266414.2 | 170 | UTR 5 | XP_005266471.1 | ||
XM_017020622.1 | 170 | Intron | XP_016876111.1 | ||
XM_017020623.1 | 170 | UTR 5 | XP_016876112.1 |