Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301025.1 | Intron | NP_001287954.1 | |||
NM_001301026.1 | Intron | NP_001287955.1 | |||
NM_003257.4 | Intron | NP_003248.3 | |||
NM_175610.3 | Intron | NP_783297.2 | |||
XM_005254617.3 | Intron | XP_005254674.2 | |||
XM_005254618.3 | Intron | XP_005254675.1 | |||
XM_005254619.3 | Intron | XP_005254676.2 | |||
XM_005254620.3 | Intron | XP_005254677.2 | |||
XM_011521972.2 | Intron | XP_011520274.2 | |||
XM_017022521.1 | Intron | XP_016878010.1 | |||
XM_017022522.1 | Intron | XP_016878011.1 | |||
XM_017022523.1 | Intron | XP_016878012.1 | |||
XM_017022524.1 | Intron | XP_016878013.1 | |||
XM_017022525.1 | Intron | XP_016878014.1 | |||
XM_017022526.1 | Intron | XP_016878015.1 | |||
XM_017022527.1 | Intron | XP_016878016.1 | |||
XM_017022528.1 | Intron | XP_016878017.1 | |||
XM_017022529.1 | Intron | XP_016878018.1 | |||
XM_017022530.1 | Intron | XP_016878019.1 | |||
XM_017022531.1 | Intron | XP_016878020.1 |