Product Details

SNP ID
rs534051
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:166611999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGCACACACACATGCACATGTGC[A/G]AGCATACACACACACACACACACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FMO9P PubMed Links

Gene Details

Gene
FMO9P
Gene Name
flavin containing monooxygenase 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details