Product Details

SNP ID
rs765427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109014309 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGCCAGGCACAGAGCAGTTTCTT[C/T]ATAACTGTTCATTGCATGAAAACAC
Phenotype
MIM: 604785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286974.1 Intron NP_001273903.1
NM_003798.3 Intron NP_003789.1
XM_005252291.3 Intron XP_005252348.1
XM_017015250.1 Intron XP_016870739.1
XM_017015251.1 Intron XP_016870740.1
Gene
TMEM245
Gene Name
transmembrane protein 245
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032012.3 Intron NP_114401.2
XM_011518446.2 Intron XP_011516748.1
XM_011518447.1 Intron XP_011516749.1
XM_011518449.1 Intron XP_011516751.1
XM_011518451.1 Intron XP_011516753.1
XM_011518452.1 Intron XP_011516754.1
XM_017014571.1 Intron XP_016870060.1
XM_017014572.1 Intron XP_016870061.1

View Full Product Details