Product Details

SNP ID
rs575710
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:29120143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACCTAAACTCAAGTCATTTTCT[A/G]AAGTAAGTTACCCACATTGACCAAA
Phenotype
MIM: 130500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166005.1 1760 Intron NP_001159477.1
NM_001166006.1 1760 Intron NP_001159478.1
NM_001166007.1 1760 Intron NP_001159479.1
NM_004437.3 1760 Intron NP_004428.1
NM_203342.2 1760 Intron NP_976217.1
NM_203343.2 1760 Intron NP_976218.1
XM_005245753.1 1760 Intron XP_005245810.1
XM_005245757.1 1760 Intron XP_005245814.1
XM_005245760.1 1760 Intron XP_005245817.1
XM_005245761.1 1760 Intron XP_005245818.1
XM_005245763.1 1760 Intron XP_005245820.1
XM_005245764.1 1760 Intron XP_005245821.1
XM_005245765.1 1760 Intron XP_005245822.1
XM_005245768.1 1760 Intron XP_005245825.1
XM_005245769.1 1760 Intron XP_005245826.1
XM_005245770.1 1760 Intron XP_005245827.1
XM_005245772.4 1760 Intron XP_005245829.1
XM_005245773.4 1760 Intron XP_005245830.1
XM_005245774.1 1760 Intron XP_005245831.1
XM_006710434.1 1760 Intron XP_006710497.1
XM_006710439.1 1760 Intron XP_006710502.1
XM_011540956.1 1760 Intron XP_011539258.1
XM_011540957.1 1760 Intron XP_011539259.1
XM_011540958.1 1760 Intron XP_011539260.1
XM_011540959.1 1760 Intron XP_011539261.1
XM_011540960.1 1760 Intron XP_011539262.1
XM_011540961.1 1760 Intron XP_011539263.1
XM_011540962.1 1760 Intron XP_011539264.1
XM_011540963.2 1760 Intron XP_011539265.1
XM_011540964.1 1760 Intron XP_011539266.1
XM_011540965.2 1760 Intron XP_011539267.1
XM_017000581.1 1760 Intron XP_016856070.1
XM_017000582.1 1760 Intron XP_016856071.1
XM_017000583.1 1760 Intron XP_016856072.1
XM_017000584.1 1760 Intron XP_016856073.1
XM_017000585.1 1760 Intron XP_016856074.1
XM_017000586.1 1760 Intron XP_016856075.1
XM_017000587.1 1760 Intron XP_016856076.1
XM_017000588.1 1760 Intron XP_016856077.1
XM_017000589.1 1760 Intron XP_016856078.1
XM_017000590.1 1760 Intron XP_016856079.1
XM_017000591.1 1760 Intron XP_016856080.1
XM_017000592.1 1760 Intron XP_016856081.1
XM_017000593.1 1760 Intron XP_016856082.1
XM_017000594.1 1760 Intron XP_016856083.1
XM_017000595.1 1760 Intron XP_016856084.1
XM_017000596.1 1760 Intron XP_016856085.1
XM_017000597.1 1760 Intron XP_016856086.1
XM_017000598.1 1760 Intron XP_016856087.1
XM_017000599.1 1760 Intron XP_016856088.1
XM_017000600.1 1760 Intron XP_016856089.1
XM_017000601.1 1760 Intron XP_016856090.1
XM_017000602.1 1760 Intron XP_016856091.1
XM_017000603.1 1760 Intron XP_016856092.1
XM_017000604.1 1760 Intron XP_016856093.1
Gene
TMEM200B
Gene Name
transmembrane protein 200B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003682.3 1760 UTR 3 NP_001003682.1
NM_001171868.1 1760 UTR 3 NP_001165339.1
XM_011541457.2 1760 UTR 3 XP_011539759.1

View Full Product Details