Product Details
- SNP ID
-
rs472509
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:86922197 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTTTTAAGTGGTTGCTTAAGGG[C/T]TTATGGTATACATGATAGAATTTCA
- Phenotype
-
MIM: 604844
MIM: 606254
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HS2ST1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141078792] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HS2ST1
- Gene Name
- heparan sulfate 2-O-sulfotransferase 1
- Gene
- SEP15
- Gene Name
- 15 kDa selenoprotein
There are no transcripts associated with this gene.
View Full Product Details